Phenylmaleic anhydride 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Phenylfuran-2,5-dione, 3-Phenyl-2,5-furandione
310400010
310400100
ACRO310400010EA
27.4
EUR
ACRO310400010
ACRO310400100
Phenylmaleic anhydride 99%
Phenylmaleic anhydride
Formel:
C₁₀H₆O₃ Molecular Weight: 174,16 g/mol Siedepunkt: 122…125 °C (,2Torr) Schmelzpunkt: 119…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015472 CAS-Nummer: 36122-35-7 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder or needles |
Infrared spectrum | Conforms |
Melting point | 119 to 123 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...