Ammonium iron(III) oxalate trihydrate 98%, rein
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Iron ammonium oxalate, Ammonium ferric oxalate, Ammonium iron oxalate trihydrate
335100500
335100010
335100051
ACRO335100500EA
34.1
EUR
ACRO335100500
ACRO335100010
ACRO335100051
Ammonium iron(III) oxalate trihydrate 98%, rein
Ammonium iron(III) oxalate trihydrate
Formel:
(NH₄)₃Fe(C₂O₄)₃·3H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150137 CAS-Nummer: 13268-42-3 UN: 2923 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Green Crystalline powder |
Infrared spectrum | Conforms |
Titration with KMnO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...