Methyl-α-D-galactopyranoside (aus Rind) 98+%

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, Methyl α-D-galactoside

337090025 337090100 337090500
ACRO337090025EA 60.7 EUR
ACRO337090025 ACRO337090100 ACRO337090500
Methyl-α-D-galactopyranoside (aus Rind) 98+%
Methyl-α-D-galactopyranoside

EINECS 222-251-7 Solubility in water: soluble Solubility in other solvents: soluble in methanol Origin bovine (cattle)


Solubility in water: soluble
Solubility in other solvents: soluble in methanol

Formel: C₇H₁₄O₆
Molecular Weight: 194.18 g/mol
Schmelzpunkt: 116…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00064085
CAS-Nummer: 3396-99-4
EINECS: 222-251-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water 7.5 to 9.5 % (K.F.)
HPLC ≥98.0 % (ELSD)
Appearance of solution (5% in water) Clear colorless
Specific optical rotation +175° to +181° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR