4-Methylumbelliferyl-α-D-galactopyranoside hydrate 99%

Lieferant: Thermo Scientific

Synonyme: ((β-D)-galactosyloxy)-isomer, 4-methylumbelliferyl galactose, 4-methylumbelliferone-α-D-galactoside, 4-methylumbelliferyl-galactopyranoside, 4-MU-α-D-Gal

337160500 337162500
ACRO337160500EA 171 EUR
ACRO337160500 ACRO337162500
4-Methylumbelliferyl-α-D-galactopyranoside hydrate 99%
4-Methylumbelliferyl-alpha-D-galactopyranoside hydrate
Formel: C₁₆H₁₈O₈.xH₂O
Molecular Weight: 338,312 g/mol (anhy)
Schmelzpunkt: 171…173 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03791284
CAS-Nummer: 38597-12-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
HPLC ≥98.5 %
Appearance of solution (5% soln. in DMF) clear, practically colorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR