(R)-2-Amino-2-phenylacetamide 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
D(-)-Phenylglycinamide, H-D-Phg-NH₂·HCl, H-D-Phg-NH2
346700050
346700250
ACRO346700050EA
84
EUR
ACRO346700050
ACRO346700250
(R)-2-Amino-2-phenylacetamide 99%
(R)-2-Amino-2-phenylacetamide
Formel:
C₈H₁₀N₂O Molecular Weight: 150,181 g/mol Schmelzpunkt: 125…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06799064 CAS-Nummer: 6485-67-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Specific optical rotation | -110° to -120° (20°C, 589 nm) (c=0.55, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...