2,5-Diaminopyridin 98+%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Pyridin-2,5-diyldiamin
360690010
360690050
360690250
ACRO360690010EA
77
EUR
ACRO360690010
ACRO360690050
ACRO360690250
2,5-Diaminopyridin 98+%
2,5-Diaminopyridin
Formel:
C₅H₇N₃ Molecular Weight: 109,13 g/mol Siedepunkt: 180 °C (12 mmHg) Schmelzpunkt: 110,3 °C Dichte: 1,251 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129023 CAS-Nummer: 4318-76-7 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Yellow to purple Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 106 to 110 °C |
Additional info | May contain up to about 10% water |
HPLC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...