Sodium pivalate hydrate 99%
Lieferant: Thermo Scientific
Achtung
Synonyme: Trimethylacetic acid sodium salt hydrate, Sodium trimethylacetate hydrate, Propanoic acid, 2,2-dimethyl-, sodium salt, hydrate
364710100 364710500
ACRO364710100EA 75.7 EUR
ACRO364710100 ACRO364710500
Sodium pivalate hydrate 99%
Sodium pivalate hydrate
Formel: C₅H₁₁NaO₃
Molecular Weight: 142,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150782
CAS-Nummer: 143174-36-1
Bestellen
Spezifikation Testergebnisse
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤7 % (K.F.)
Titration with HClO4 ≥98.5 % (On dry substance)