L-tert-Leucinol 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-tert-Leucinol, (S)-2-Amino-3,3-dimethylbutan-1-ol, (S)-2-Amino-3,3-dimethylbutanol
377530010
377530050
ACRO377530010EA
82.1
EUR
ACRO377530010
ACRO377530050
L-tert-Leucinol 95%
L-tert-Leucinol
Formel:
C₆H₁₅NO Molecular Weight: 117,19 g/mol Siedepunkt: 114…116 °C (10 mmHg) Schmelzpunkt: 30…34 °C Flammpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192250 CAS-Nummer: 112245-13-3 |
Spezifikation Testergebnisse
Appearance | Colorless to white Low melting mass or liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Specific optical rotation | +34° to +40° (25°C, 589 nm) (c=1.5, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...