4-Amino-1-Boc-piperidin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester
384860010
384860050
ACRO384860010EA
22.3
EUR
ACRO384860010
ACRO384860050
4-Amino-1-Boc-piperidin 98%
4-Amino-1-Boc-piperidin
Formel:
C₁₀H₂₀N₂O₂ Molecular Weight: 200,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01076201 CAS-Nummer: 87120-72-7 UN: 3259 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to light yellow to beige Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...