4-(Aminomethyl)tetrahydropyran 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine
385210010
385210050
385210250
ACRO385210010EA
115
EUR
ACRO385210010
ACRO385210050
ACRO385210250
4-(Aminomethyl)tetrahydropyran 97%
4-(Aminomethyl)tetrahydropyran
Formel:
C₆H₁₃NO Molecular Weight: 115,1735 g/mol Siedepunkt: 75…76 °C (18 mmHg) Dichte: 1,02 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179435 CAS-Nummer: 130290-79-8 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4640 to 1.4670 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...