Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal®
Lieferant: Thermo Scientific
Gefahr
Synonyme: Isopropylmagnesium chloride - Lithium chloride complex, Turbo Grignard
386281000 386288000
386281000.EA 122 EUR
386281000. 386288000.
Isopropylmagnesium chloride - LiCl complex 1.3 M in THF, AcroSeal®
Isopropylmagnesium chloride - LiCl complex
Formel: C₃H₇ClMg·LiCl
Molecular Weight: 145,24 g/mol
Dichte: 0,98 g/cm³
Flammpunkt: –21 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784514
CAS-Nummer: 745038-86-2
UN: 3399
ADR: 4.3,I
Bestellen
Spezifikation Testergebnisse
Appearance Clear yellow to grey to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.1 to 1.5 M