(S)-N-Boc-allylglycine 95%
Lieferant: Thermo Scientific
Synonyme:
(S)-2-((tert-Butoxycarbonyl)amino)pent-4-enoic acid, Boc-Allyl-Gly-OH, (S)-2-Boc-AMino-4-pentenoic acid, 4-Pentenoic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, (2S)-
390100010
390105000
390100050
ACRO390100010EA
194
EUR
ACRO390100010
ACRO390105000
ACRO390100050
(S)-N-Boc-allylglycine 95%
(S)-N-Boc-allylglycine
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215,249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320851 CAS-Nummer: 90600-20-7 |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Viscous liquid or paste |
Infrared spectrum | Conforms |
Specific optical rotation | +9.0° to +13.0° (20°C, 589 nm) (c=1, methanol) |
Titration with NaOH | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...