4'-Ethynylacetophenone 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(4-Ethynyl-phenyl)-ethanone, 1-(4-Ethynylphenyl)ethanone, 1 - (4 - Ethynyl - phenyl) - ethanone, Ethanone,1-(4-ethynylphenyl)-
390580010
390580050
ACRO390580010EA
129
EUR
ACRO390580010
ACRO390580050
4'-Ethynylacetophenone 98%
4'-Ethynylacetophenone
Formel:
C₁₀H₈O Molecular Weight: 144,173 g/mol Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974025 CAS-Nummer: 42472-69-5 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Powder |
Infrared spectrum | Conforms |
Melting point | 67 to 70 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...