3-Iodbenzaldehyd 99%

Lieferant: Thermo Scientific

392070010 392070050
ACRO392070010EA 79 EUR
ACRO392070010 ACRO392070050
3-Iodbenzaldehyd 99%
3-Iodobenzaldehyde
Formel: C₇H₅IO
Molecular Weight: 232,02 g/mol
Schmelzpunkt: 57…60 °C
MDL: MFCD00039573
CAS-Nummer: 696-41-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR