Methyl-6-bromhexanoat 96+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Bromo-hexanoic acid methyl ester
392690050
392690250
392691000
ACRO392690050EA
25.6
EUR
ACRO392690050
ACRO392690250
ACRO392691000
Methyl-6-bromhexanoat 96+%
Methyl-6-bromhexanoat
Formel:
C₇H₁₃BrO₂ Molecular Weight: 209.084 g/mol Siedepunkt: 150 °C (50 mmHg) Dichte: 1.316 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00187528 CAS-Nummer: 14273-90-6 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4630 to 1.4650 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...