(2-Iodphenyl)essigsäure 97+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(2-Iodophenyl)acetic acid

394040010 394040050 394040250
ACRO394040010EA 19.6 EUR
ACRO394040010 ACRO394040050 ACRO394040250
(2-Iodphenyl)essigsäure 97+%
(2-Iodphenyl)essigsäure
Formel: (C₆H₄I)CH₂COOH
Molecular Weight: 262.05 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00046546
CAS-Nummer: 18698-96-9

Bestellen

Spezifikation Testergebnisse

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 113 to 118 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR