2-Phenylthiazol 98%
Lieferant: Thermo Scientific
|
Achtung
|
394370010
394370050
ACRO394370010EA
50.7
EUR
ACRO394370010
ACRO394370050
2-Phenylthiazol 98%
2-Phenylthiazole
Formel:
C₉H₇NS Molecular Weight: 161,2200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272327 CAS-Nummer: 1826-11-5 |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...