4-Biphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
[1,1'-Biphenyl]-4-ylboronic acid
394420010
394420050
394420250
ACRO394420010EA
34.2
EUR
ACRO394420010
ACRO394420050
ACRO394420250
4-Biphenylboronsäure 97%
4-Biphenylboronsäure
Formel:
C₁₂H₁₁BO₂ Molecular Weight: 198,03 g/mol Schmelzpunkt: 241…245 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093311 CAS-Nummer: 5122-94-1 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...