(S)-(-)-N,α-Dimethylbenzylamin 99+%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (S)-N,α-Dimethylbenzylamine, (S)-(-)-alpha-Methyl-N-methylbenzylamine, (S)-(-)-N-Methyl-1-phenylethylamine, (S)-(-)-N α-Dimethylbenzylamine

395840010 395840050
ACRO395840010EA 47.7 EUR
ACRO395840010 ACRO395840050
(S)-(-)-N,α-Dimethylbenzylamin 99+%
(S)-(-)-N,α-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135,2100 g/mol
Siedepunkt: 74…76 °C (11 mmHg)
Dichte: 0,924 g/cm³
Flammpunkt: 61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067113
CAS-Nummer: 19131-99-8
UN: 3267
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5130 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation -63° to -67° (20°C, 589 nm) (c=4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR