2,6-Dichlornicotinsäure 98+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,6-Dichlor-3-pyridincarbonsäure, 2,6-Dichlorpyridin-3-carbonsäure

396610010 396610050 396610250
ACRO396610010EA 30 EUR
ACRO396610010 ACRO396610050 ACRO396610250
2,6-Dichlornicotinsäure 98+%
2,6-Dichlornicotinsäure
Formel: C₆H₃Cl₂NO₂
Schmelzpunkt: 147…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075583
CAS-Nummer: 38496-18-3

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR