MES Natriumsalz 99%
Lieferant: Thermo Scientific
Synonyme:
MES Na, Natrium-3-morpholin-1-ylethylsulfonat, 2-(N-Morpholino)ethansulfonsäure Natriumsalz
397350250
397351000
397355000
ACRO397350250EA
55.3
EUR
ACRO397350250
ACRO397351000
ACRO397355000
MES Natriumsalz 99%
MES Natriumsalz
Formel:
C₆H₁₂NNaO₄S Molecular Weight: 217,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065473 CAS-Nummer: 71119-23-8 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Solubility | (10 % in water) Clear colorless |
UV | at 280 nm A: ≤0.03 |
Titration after Ion exchange | ≥98.5 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...