4-Aminoimidazole-5-carboxamide 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-1H-imidazole-5-carboxamide, 5-Aminoimidazole-4-carboxamide, 5-Amino-4-imidazolecarboxamide, 5-Amino-1H-imidazole-4-carboxamide, 4-Aminoimidazole-5-carboxamide, 4-aminoimidazole-5-carboxamide, 5-Amino-3H-imidazole-4-Carboxamide

397550050 397550250
ACRO397550050EA 72.4 EUR
ACRO397550050 ACRO397550250
4-Aminoimidazole-5-carboxamide 95%
4-Aminoimidazole-5-carboxamide
Formel: C₄H₆N₄O
Molecular Weight: 126,12 g/mol
Schmelzpunkt: 164…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02181040
CAS-Nummer: 360-97-4

Bestellen

Spezifikation Testergebnisse

Appearance Grey to purple Powder
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR