5-Fluorsalicylaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Fluoro-2-hydroxybenzaldehyde

397570010 397570050
ACRO397570010EA 107 EUR
ACRO397570010 ACRO397570050
5-Fluorsalicylaldehyd 97%
5-Fluorsalicylaldehyd
Formel: C₇H₅FO₂
Molecular Weight: 140,11 g/mol
Schmelzpunkt: 82…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01090997
CAS-Nummer: 347-54-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 81 to 86 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR