(2S)-1-{[(2-Methyl-2-propanyl)oxy]carbonyl}-2-piperidincarbonsäure 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: Boc-Homopro-OH, 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, Boc-HomoPro-OH, 1-Boc-L-pipecolic Acid, (S)-1-Boc-2-piperidinecarboxylic Acid, Boc-Pip-OH, (S)-N-BOC-Piperidine-2-carboxylic acid
397830050 397830250
ACRO397830050EA 117 EUR
ACRO397830050 ACRO397830250
(2S)-1-{[(2-Methyl-2-propanyl)oxy]carbonyl}-2-piperidincarbonsäure 98%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,28 g/mol
Schmelzpunkt: 123…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151904
CAS-Nummer: 26250-84-0
Bestellen
Spezifikation Testergebnisse
Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -62° to -65° (20°C, 589 nm) (C=1, acetic acid)
Titration with NaOH ≥97.5 %