3-Cyan-4-fluorbenzaldehyd 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Fluoro-5-formylbenzonitrile

397940010 397940050
ACRO397940010EA 43.8 EUR
ACRO397940010 ACRO397940050
3-Cyan-4-fluorbenzaldehyd 99%
3-Cyano-4-fluorobenzaldehyde
Formel: C₈H₄FNO
Molecular Weight: 149,1219 g/mol
Schmelzpunkt: 80…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863558
CAS-Nummer: 218301-22-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light orange Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 89 °C
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR