Piperidin-3-amine dihydrochloride 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: piperidin-3-amine dihydrochloride, 3-Aminopiperidine dihydrochloride, 3-Amino-Piperidine Di-Hydrochloride

398110010 398112500
ACRO398110010EA 294 EUR
ACRO398110010 ACRO398112500
Piperidin-3-amine dihydrochloride 97%
Piperidin-3-amine dihydrochloride
Formel: C₅H₁₄Cl₂N₂
Molecular Weight: 173,081 g/mol
Schmelzpunkt: 230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012773
CAS-Nummer: 138060-07-8

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Powder or solid
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR