Isocytosin 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-4(1H)-pyrimidinone, 2-Aminopyrimidin-4(1H)-one

398320050 398320250
ACRO398320050EA 130 EUR
ACRO398320050 ACRO398320250
Isocytosin 99%
Isocytosine
Formel: C₄H₅N₃O
Molecular Weight: 111.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00057557
CAS-Nummer: 108-53-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Solubility (50 mg/ml in DMSO) Clear solution
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR