4-Amino-2,6-dichlorpyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,6-Dichlor-4-pyridylamin, 2,6-Dichlorpyridin-4-amin
398700010
398700050
ACRO398700010EA
24
EUR
ACRO398700010
ACRO398700050
4-Amino-2,6-dichlorpyridin 98%
4-Amino-2,6-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,01 g/mol Schmelzpunkt: 167…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052832 CAS-Nummer: 2587-02-2 |
Spezifikation Testergebnisse
Appearance | White to yellow-brown Powder |
Infrared spectrum | Conforms |
Melting point | 169 to 173 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...