D-Valin-methylester-hydrochlorid 99%
Lieferant: Thermo Scientific
Synonyme: H-D-Val-OMe.HCl, H-D-Val-OMe·HCl
398720010 398720050
ACRO398720010EA 25.5 EUR
ACRO398720010 ACRO398720050
D-Valin-methylester-hydrochlorid 99%
D-Valine methyl ester hydrochloride
Formel: C₆H₁₃NO₂
Molecular Weight: 167,633 g/mol
Schmelzpunkt: 158…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00237309
CAS-Nummer: 7146-15-8
Bestellen
Spezifikation Testergebnisse
Appearance White Powder
Infrared spectrum Conforms
Specific optical rotation -13.5° to -17.5° (20°C, 589 nm) (c=2,H2O)
Titration with HClO4 ≥98.5 %