5-Bromoxindol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-1,3-dihydro-2H-indol-2-one, 5-Bromoindolin-2-one, 5-Bromo-2-oxindole

398530010 398530050
ACRO398530010EA 35 EUR
ACRO398530010 ACRO398530050
5-Bromoxindol 97%
5-Bromoxindol
Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00456998
CAS-Nummer: 20870-78-4

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to tan Powder or crystals
Infrared spectrum Conforms
Melting point 220 to 224 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR