Tetrakis(acetonitrile)copper(I) tetrafluoroborate

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Copper(I) tetra(acetonitrile) tetrafluoroborate, Tetrakis(acetonitrile)copper(I) tetrafluoroborate

398760010 398760050
ACRO398760010EA 30.1 EUR
ACRO398760010 ACRO398760050
Tetrakis(acetonitrile)copper(I) tetrafluoroborate
Tetrakis(acetonitrile)copper(I) tetrafluoroborate
Formel: C₈H₁₂BCuF₄N₄
Molecular Weight: 314.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265110
CAS-Nummer: 15418-29-8
UN: 1759
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 19.5 to 20.5 % Cu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR