N-Boc-S-trityl-L-cysteine 99%

Lieferant: Thermo Scientific

Synonyme: N-Boc-S-trityl-L-cysteine, N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, BOC-S-trityl-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tritylthio)propanoic acid, Boc-Cys(Trt)-OH

399060010 399060050
ACRO399060010EA 26.5 EUR
ACRO399060010 ACRO399060050
N-Boc-S-trityl-L-cysteine 99%
N-Boc-S-trityl-L-cysteine
Formel: C₂₇H₂₉NO₄S
Molecular Weight: 463,592 g/mol
Schmelzpunkt: 143…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00038251
CAS-Nummer: 21947-98-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 142 to 154 °C
HPLC ≥98.5 %
Specific optical rotation +26° to +30° (20 °C, 589 nm) (c=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR