Peanut oil
Lieferant: Thermo Fisher Scientific
416855000
416850025
ACRO416855000EA
88.1
EUR
ACRO416855000
ACRO416850025
Peanut oil
Peanut oil
Dichte:
0.915 g/cm³ Flammpunkt: 283 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00131805 CAS-Nummer: 8002-03-7 |
Spezifikation Testergebnisse
Appearance | Clear yellow Oily liquid |
Infrared spectrum | Conforms |
Water | ≤0.1 % |
Composition of fatty acids | 0.5 to 3.0 % (GC) Lignoceric acid |
Acid value | ≤0.5 mg KOH/g |
Peroxide value | ≤5 meq O2/kg |
Unsaponifiable matter | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...