4-Methyl-1H-imidazole-5-carbaldehyde 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyl-5-imidazolecarboxaldehyde

425770050 425770250
ACRO425770050EA 67.7 EUR
ACRO425770050 ACRO425770250
4-Methyl-1H-imidazole-5-carbaldehyde 99%
4-Methyl-1H-imidazole-5-carbaldehyde
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 165…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173728
CAS-Nummer: 68282-53-1

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 170 °C
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR