L-β-Homoproline hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-2-(Pyrrolidin-2-yl)acetic acid hydrochloride, H-β-HoPro-OH·HCl

426170010 426172500
ACRO426170010EA 420 EUR
ACRO426170010 ACRO426172500
L-β-Homoproline hydrochloride 98%
L-beta-Homoproline hydrochloride
Formel: C₆H₁₂ClNO₂
Molecular Weight: 165.617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07363484
CAS-Nummer: 53912-85-9

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR