2-Indancarboxylic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,3-Dihydro-1H-indene-2-carboxylic acid, Indan-2-carboxylic acid

426360010 426360050
ACRO426360010EA 117 EUR
ACRO426360010 ACRO426360050
2-Indancarboxylic acid 98%
2-Indancarboxylic acid
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
Schmelzpunkt: 128…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085095
CAS-Nummer: 25177-85-9

Bestellen

Spezifikation Testergebnisse

Appearance White to cream to brown Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Sulfated ash ≤0.1 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR