3-(Benzyldimethylammonio)propanesulphonate 95%

Lieferant: Thermo Scientific

Synonyme: NDSB-256, 3-(Benzyldimethylammonio) propane-1-sulphonate, 3-[dimethylbenzylamino]propanesulphonic acid

428070050 428070250
ACRO428070050EA 102 EUR
ACRO428070050 ACRO428070250
3-(Benzyldimethylammonio)propanesulphonate 95%
3-(Benzyldimethylammonio)propanesulphonate
Formel: C₁₂H₁₉NO₃S
Molecular Weight: 257,348 g/mol
Schmelzpunkt: 245…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00225018
CAS-Nummer: 81239-45-4

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
NMR ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR