Butvar® B-98

Lieferant: Thermo Scientific

Synonyme: Polyvinyl butyral resin

428482500 428485000
ACRO428482500EA 96.3 EUR
ACRO428482500 ACRO428485000
Butvar® B-98
Butvar® B-98
Dichte: 1,1 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00084458
CAS-Nummer: 63148-65-2

Bestellen

Spezifikation Testergebnisse

Appearance White to yellowish-white Powder
Infrared spectrum Conforms
Assay ≤2.5 % (acetate content) as PV acetate
Molecular weight 40000 to 70000 (by SEC, average)
Viscosity 6 to 9 cP (6 % in methanol at 20°C)
Volatile matter ≤5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR