3-Brombenzylamin 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(3-Bromophenyl)methanamine
428960010
428960050
428960250
ACRO428960010EA
27.2
EUR
ACRO428960010
ACRO428960050
ACRO428960250
3-Brombenzylamin 95%
3-Brombenzylamin
Formel:
C₇H₈BrN Molecular Weight: 186,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01026119 CAS-Nummer: 10269-01-9 UN: 2735 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5850 to 1.5900 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...