6-Methoxypyridine-3-sulphonyl chloride 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Methoxy-pyridine-3-sulphonyl chloride, 3-Pyridinesulphonylchloride, 6-methoxy-, 3-(Chlorosulphonyl)-6-methoxypyridine
429410010
429410050
ACRO429410010EA
134
EUR
ACRO429410010
ACRO429410050
6-Methoxypyridine-3-sulphonyl chloride 97%
6-Methoxypyridine-3-sulphonyl chloride
Formel:
C₆H₆ClNO₃S Molecular Weight: 207,628 g/mol Schmelzpunkt: 42…45 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD06739108 CAS-Nummer: 312300-42-8 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to off-white Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 42 to 46 °C |
GC | ≥96.0 % |
Absorbance | ≤0.01 (water, 1 cm, 254 nm) |
Chloride (Cl) | ≤0.1 ppm |
Nitrate (NO3) | ≤0.2 ppm |
Phosphate (PO4) | ≤0.05 ppm |
Sulfate (SO4) | ≤1 ppm |
Silicate (as SiO2) | ≤0.01 ppm |
Aluminium (Al) | ≤0.02 ppm |
Ammonium (NH4) | ≤0.01 ppm |
Antimony (Sb) | ≤0.01 ppm |
Barium (Ba) | ≤0.01 ppm |
Bismuth (Bi) | ≤0.01 ppm |
Boron (B) | ≤0.01 ppm |
Cadmium (Cd) | ≤0.01 ppm |
Calcium (Ca) | ≤0.1 ppm |
Chromium (Cr) | ≤0.01 ppm |
Cobalt (Co) | ≤0.01 ppm |
Copper (Cu) | ≤0.01 ppm |
Gold (Au) | ≤0.01 ppm |
Iron (Fe) | ≤0.01 ppm |
Lithium (Li) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Manganese (Mn) | ≤0.01 ppm |
Mercury (Hg) | ≤0.05 ppm |
Molybdenum (Mo) | ≤0.01 ppm |
Nickel (Ni) | ≤0.01 ppm |
Potassium (K) | ≤0.05 ppm |
Selenium (Se) | ≤0.01 ppm |
Silver (Ag) | ≤0.01 ppm |
Sodium (Na) | ≤0.1 ppm |
Strontium (Sr) | ≤0.05 ppm |
Tin (Sn) | ≤0.01 ppm |
Titanium (Ti) | ≤0.01 ppm |
Vanadium (V) | ≤0.01 ppm |
Zinc (Zn) | ≤0.05 ppm |
Zirconium (Zr) | ≤0.01 ppm |
Heavy metals (as Pb) | ≤0.01 ppm |
KMnO4-reducing substance | Passes test |
Conductivity | ≤6 µS/cm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...