2-Chloropyrimidine-5-carboxylic acid 97%

Lieferant: Thermo Scientific
Achtung

429470010 429470050
ACRO429470010EA 166 EUR
ACRO429470010 ACRO429470050
2-Chloropyrimidine-5-carboxylic acid 97%
2-Chloropyrimidine-5-carboxylic acid
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158,5400 g/mol
Schmelzpunkt: 125…127,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06739036
CAS-Nummer: 374068-01-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR