2-Amino-4-thiazolcarbonsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Aminothiazole-4-carboxylic acid
429480010
429480050
ACRO429480010EA
52.9
EUR
ACRO429480010
ACRO429480050
2-Amino-4-thiazolcarbonsäure 97%
2-Amino-4-thiazolecarboxylic acid
Formel:
C₄H₄N₂O₂S Molecular Weight: 144,15 g/mol Schmelzpunkt: 238…239 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00859429 CAS-Nummer: 40283-41-8 |
Spezifikation Testergebnisse
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Water | ≤0.1 % (Coulometric) |
GC | ≤0.1 % 2-Propanol |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Acidity (CH3COOH) | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...