1H-Benzimidazol-4-carbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1H-Benzimidazole-7-carboxylic acid

429510010 429510050
ACRO429510010EA 265 EUR
ACRO429510010 ACRO429510050
1H-Benzimidazol-4-carbonsäure 97%
1H-Benzimidazol-4-carbonsäure
Formel: C₈H₆N₂O₂
Molecular Weight: 162.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01823426
CAS-Nummer: 46006-36-4
UN: 3077
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR