5-Bromthiazol 95%
Lieferant: Thermo Scientific
|
Achtung
|
429980010
429980050
ACRO429980010EA
85.8
EUR
ACRO429980010
ACRO429980050
5-Bromthiazol 95%
5-Bromothiazole
Formel:
C₃H₂BrNS Molecular Weight: 164,03 g/mol Flammpunkt: 76,7 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD07787394 CAS-Nummer: 3034-55-7 |
Spezifikation Testergebnisse
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...