4-Fluor-2-(trifluormethyl)phenylboronsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Fluoro-2-(trifluoromethyl)benzeneboronic acid, (4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid
430440010
430440050
ACRO430440010EA
77.7
EUR
ACRO430440010
ACRO430440050
4-Fluor-2-(trifluormethyl)phenylboronsäure 97%
4-Fluor-2-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207.92 g/mol Schmelzpunkt: 184…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062379 CAS-Nummer: 182344-16-7 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 182 to 187 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...