3-Fluor-5-(trifluormethyl)phenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Fluoro-5-(trifluoromethyl)benzeneboronic acid, (3-Fluoro-5-(trifluoromethyl)phenyl)boronic acid, 3-(Trifluoromethyl)-5-fluorophenylboronic acid

430320010 430320050
ACRO430320010EA 155 EUR
ACRO430320010 ACRO430320050
3-Fluor-5-(trifluormethyl)phenylboronsäure 97%
3-Fluor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
Molecular Weight: 207,92 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06657735
CAS-Nummer: 159020-59-4

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 175 to 180 °C
HPLC ≥96.0 %
Substance darkened by H2SO4 Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR