3-(Trifluormethyl)-5-pyridinboronsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(5-(Trifluoromethyl)pyridin-3-yl)boronic acid, (5-Trifluoromethylpyridin-3-yl)boronic acid
430520010
430522500
ACRO430520010EA
540
EUR
ACRO430520010
ACRO430522500
3-(Trifluormethyl)-5-pyridinboronsäure 95%
3-(Trifluormethyl)-5-pyridinboronsäure
Formel:
C₆H₅BF₃NO₂ Molecular Weight: 190.9156 g/mol Schmelzpunkt: 260…262.6 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09952041 CAS-Nummer: 947533-51-9 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...