3-Chlor-4-methylphenylboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid
430620010
430620100
ACRO430620010EA
33.5
EUR
ACRO430620010
ACRO430620100
3-Chlor-4-methylphenylboronsäure 97%
3-Chlor-4-methylphenylboronsäure
Formel:
C₇H₈BClO₂ Molecular Weight: 170,40 g/mol Schmelzpunkt: 228,3…231.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04039010 CAS-Nummer: 175883-63-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...