1H-Indazole-4-carbaldehyde 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Formylindazole, 1H-Indazole-4-carboxaldehyde, 4-Formyl-1H-indazole

431030010 431030050
ACRO431030010EA 233 EUR
ACRO431030010 ACRO431030050
1H-Indazole-4-carbaldehyde 97%
1H-Indazole-4-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Schmelzpunkt: 166…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738279
CAS-Nummer: 669050-70-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
HPLC ≥96.0 %
Water adsorption ≥20 % (50% rH, 1 bar, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR