Ethyl-6-chloro-3-pyridazinecarboxylate 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 6-chloro-3-pyridazinecarboxylate, Ethyl 6-chloropyridazine-3-carboxylate, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, 6-Chloro-pyridazine-3-carboxylic acid ethyl ester

431110010 431110050
ACRO431110010EA 160 EUR
ACRO431110010 ACRO431110050
Ethyl-6-chloro-3-pyridazinecarboxylate 95%
Ethyl-6-chloro-3-pyridazinecarboxylate
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186.595 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09908168
CAS-Nummer: 75680-92-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR